missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Trigger factor Protein

A DNA sequence encoding the Escherichia coli O127:H6 (strain E2348/69 / EPEC) Trigger factor, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7443-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058880

Cantidad Precio
1 352.00€ / 10 microgramos
6 328.49€ / 10 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

WP_001198386.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
tig
Recombinant Protein
Yeast
MQVSVETTQG LGRRVTITIA ADSIETAVKS ELVNVAKKVR IDGFRKGKVP MNIVAQRYGA SVRQDVLGDL MSRNFIDAII KEKINPAGAP TYVPGEYKLG EDFTYSVEFE VYPEVELQGL EAIEVEKPIV EVTDADVDGM LDTLRKQQAT WKEKDGAVEA EDRVTIDFTG SVDGEEFEGG KASDFVLAMG QGRMIPGFED GIKGHKAGEE FTIDVTFPEE YHAENLKGKA AKFAINLKKV EERELPELTA EFIKRFGVED GSVEGLRAEV RKNMERELKS AIRNRVKSQA IEGLVKANDI DVPAALIDSE IDVLRRQAAQ RFGGNEKQAL ELPRELFEEQ AKRRVVVGLL LGEVIRTNEL KADEERVKGL IEEMASAYED PKEVIEFYSK NKELMDNMRN VALEEQAVEA VLAKAKVTEK ETTFNELMNQ QA
Trigger factor Protein
10 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
Videos
Documentos