missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli SlyD Protein
A cDNA sequence encoding the SlyD was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10865-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
947859 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 220 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-Hcl pH8.0 using chymotrypsin. | |
E. coli | |
Untagged | |
SlyD protein solution contains 20mM Tris pH 7.5. |
SlyD Protein | |
Research Use Only | |
slyD | |
Recombinant Protein | |
E. coli | |
MKVAKDLVVS LAYQVRTEDG VLVDESPVSA PLDYLHGHGS LISGLETALE GHEVGDKFDV AVGANDAYGQ YDENLVQRVP KDVFMGVDEL QVGMRFLAET DQGPVPVEIT AVEDDHVVVD GNHMLAGQNL KFNVEVVAIR EATEEELAHG HVHGAHDHHH DHDHDGCCGG HGHDHGHEHG GEGCCGGKGN GGCGCH | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |