missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Phosphoheptose isomerase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Phosphoheptose isomerase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7258-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_414757.1 | |
Phosphoheptose isomerase Protein | |
Research Use Only | |
gmhA | |
Recombinant Protein | |
E. coli | |
MYQDLIRNEL NEAAETLANF LKDDANIHAI QRAAVLLADS FKAGGKVLSC GNGGSHCDAM HFAEELTGRY RENRPGYPAI AISDVSHISC VGNDFGFNDI FSRYVEAVGR EGDVLLGIST SGNSANVIKA IAAAREKGMK VITLTGKDGG KMAGTADIEI RVPHFGYADR IQEIHIKVIH ILIQLIEKEM VK | |
Greater than 90% as determined by SDS-PAGE. |
949134 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |