missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Oxidoreductase YdhF Protein

A DNA sequence encoding the Escherichia coli (strain K12) Oxidoreductase YdhF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6940-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056780

  • 414.00€ / 100 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

WP_000250656.1
Oxidoreductase YdhF Protein
100 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
946960
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ydhF
Recombinant Protein
E. coli
MVQRITIAPQ GPEFSRFVMG YWRLMDWNMS ARQLVSFIEE HLDLGVTTVD HADIYGGYQC EAAFGEALKL APHLRERMEI VSKCGIATTA REENVIGHYI TDRDHIIKSA EQSLINLATD HLDLLLIHRP DPLMDADEVA DAFKHLHQSG KVRHFGVSNF TPAQFALLQS RLPFTLATNQ VEISPVHQPL LLDGTLDQLQ QLRVRPMAWS CLGGGRLFND DYFQPLRDEL AVVAEELNAG SIEQVVYAWV LRLPSQPLPI IGSGKIERVR AAVEAETLKM TRQQWFRIRK AALGYDVP
Videos
Documentos