missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli nrdA Protein
A DNA sequence encoding the Escherichia coli (strain K12) nrdA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7262-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_416737.1 | |
nrdA Protein | |
Research Use Only | |
nrdA | |
Recombinant Protein | |
E. coli | |
MNQNLLVTKR DGSTERINLD KIHRVLDWAA EGLHNVSISQ VELRSHIQFY DGIKTSDIHE TIIKAAADLI SRDAPDYQYL AARLAIFHLR KKAYGQFEPP ALYDHVVKMV EMGKYDNHLL EDYTEEEFKQ MDTFIDHDRD MTFSYAAVKQ LEGKYLVQNR VTGEIYESAQ FLYILVAACL FSNYPRETRL QYVKRFYDAV STFKISLPTP IMSGVRTPTR QFSSCVLIEC GDSLDSINAT SSAIVKYVSQ RAGIGINAGR IRALGSPIRG GEAFHTGCIP FYKHFQTAVK SCSQGGVRGG AATLFYPMWH LEVESLLVLK NNRGVEGNRV RHMDYGVQIN KLMYTRLLKG EDITLFSPSD VPGLYDAFFA DQEEFERLYT KYEKDDSIRK QRVKAVELFS LMMQERASTG RIYIQNVDHC NTHSPFDPAI APVRQSNLCL EIALPTKPLN DVNDENGEIA LCTLSAFNLG AINNLDELEE LAILAVRALD ALLDYQDYPI PAAKRGAMGR RTLGIGVINF AYYLAKHGKR YSDGSANNLT HKTFEAIQYY LLKASNELAK EQGACPWFNE TTYAKGILPI DTYKKDLDTI ANEPLHYDWE ALRESIKTHG LRNSTLSALM PSETSSQISN ATNGIEPPRG YVSIKASKDG ILRQVVPDYE HLHDAYELLW EMPGNDGYLQ LVGIMQKFID QSISANTNYD PSRFPSGKVP MQQLLKDLLT AYKFGVKTLY YQNTRDGAED AQDDLVPSIQ DDGCESGACK I | |
Greater than 90% as determined by SDS-PAGE. |
946612 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |