missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli Ecotin Protein

A cDNA sequence encoding the Ecotin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11741-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908208

  • 160.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

Ecotin Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
The Ecotin protein solution (1 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50mM NaCl and 10% glycerol.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAESVQPLEK IAPYPQAEKG MKRQVIQLTP QEDESTLKVE LLIGQTLEVD CNLHRLGGKL ENKTLEGWGY DYYVFDKVSS PVSTMMACPD GKKEKKFVTA YLGDAGMLRY NSKLPIVVYT PDNVDVKYRV WKAEEKIDNA VVR
Videos