missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cell division protein FtsZ Protein

A DNA sequence encoding the Escherichia coli O6:H1 (strain CFT073 / ATCC 700928 / UPEC) Cell division protein FtsZ, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7325-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058210

  • 1337.00€ / 200 microgramos


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

WP_000462776.1
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ftsZ
Recombinant Protein
E. coli
MFEPMELTND AVIKVIGVGG GGGNAVEHMV RERIEGVEFF AVNTDAQALR KTAVGQTIQI GSGITKGLGA GANPEVGRNA ADEDRDALRA ALEGADMVFI AAGMGGGTGT GAAPVVAEVA KDLGILTVAV VTKPFNFEGK KRMAFAEQGI TELSKHVDSL ITIPNDKLLK VLGRGISLLD AFGAANDVLK GAVQGIAELI TRPGLMNVDF ADVRTVMSEM GYAMMGSGVA SGEDRAEEAA EMAISSPLLE DIDLSGARGV LVNITAGFDL RLDEFETVGN TIRAFASDNA TVVIGTSLDP DMNDELRVTV VATGIGMDKR PEITLVTNKQ VQQPVMDRYQ QHGMAPLTQE QKPVAKVVND NAPQTAKEPD YLDIPAFLRK QAD
Cell division protein FtsZ Protein
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Videos
Documentos