missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Agmatinase Protein
A DNA sequence encoding the Escherichia coli (strain 55989 / EAEC) Agmatinase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5622-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
WP_000105559.1 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Agmatinase Protein | |
Research Use Only | |
speB | |
Recombinant Protein | |
E. coli | |
MSTLGHQYDN SLVSNAFGFL RLPMNFQPYD SDADWVITGV PFDMATSGRA GGRHGPAAIR QVSTNLAWEH NRFPWNFDMR ERLNVVDCGD LVYAFGDARE MSEKLQAHAE KLLAAGKRML SFGGDHFVTL PLLRAHAKHF GKMALVHFDA HTDTYANGCE FDHGTMFYTA PKEGLIDPNH SVQIGIRTEF DIDNGFTVLD ACQVNDRSVD DVIAQVKQIV GDMPVYLTFD IDCLDPAFAP GTGTPVIGGL TSDRAIKLVR GLKDLNIVGM DVVEVAPAYD QSEITALAAA TLALEMLYIQ AAKKGE | |
Greater than 90% as determined by SDS-PAGE. |