missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli ADK Protein
A DNA sequence encoding the Escherichia coli (strain K12) ADK, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8857-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_415007.1 | |
ADK Protein | |
Research Use Only | |
adk | |
Recombinant Protein | |
E. coli | |
MRIILLGAPG AGKGTQAQFI MEKYGIPQIS TGDMLRAAVK SGSELGKQAK DIMDAGKLVT DELVIALVKE RIAQEDCRNG FLLDGFPRTI PQADAMKEAG INVDYVLEFD VPDELIVDRI VGRRVHAPSG RVYHVKFNPP KVEGKDDVTG EELTTRKDDQ EETVRKRLVE YHQMTAPLIG YYSKEAEAGN TKYAKVDGTK PVAEVRADLE KILG | |
Greater than 90% as determined by SDS-PAGE. |
945097 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |