missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Adenosine deaminase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Adenosine deaminase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5611-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936429

  • 2645.00€ / 1 miligramo


Fecha estimada de envío
01-04-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_416140.1
Adenosine deaminase Protein
1 mg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
945851
Greater than 90% as determined by SDS-PAGE.
Research Use Only
add
Recombinant Protein
E. coli
MIDTTLPLTD IHRHLDGNIR PQTILELGRQ YNISLPAQSL ETLIPHVQVI ANEPDLVSFL TKLDWGVKVL ASLDACRRVA FENIEDAARH GLHYVELRFS PGYMAMAHQL PVAGVVEAVI DGVREGCRTF GVQAKLIGIM SRTFGEAACQ QELEAFLAHR DQITALDLAG DELGFPGSLF LSHFNRARDA GWHITVHAGE AAGPESIWQA IRELGAERIG HGVKAIEDRA LMDFLAEQQI GIESCLTSNI QTSTVAELAA HPLKTFLEHG IRASINTDDP GVQGVDIIHE YTVAAPAAGL SREQIRQAQI NGLEMAFLSA EEKRALREKV AAK
Videos
Documentos