missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Adenosine deaminase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Adenosine deaminase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5611-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_416140.1 | |
Adenosine deaminase Protein | |
Research Use Only | |
add | |
Recombinant Protein | |
E. coli | |
MIDTTLPLTD IHRHLDGNIR PQTILELGRQ YNISLPAQSL ETLIPHVQVI ANEPDLVSFL TKLDWGVKVL ASLDACRRVA FENIEDAARH GLHYVELRFS PGYMAMAHQL PVAGVVEAVI DGVREGCRTF GVQAKLIGIM SRTFGEAACQ QELEAFLAHR DQITALDLAG DELGFPGSLF LSHFNRARDA GWHITVHAGE AAGPESIWQA IRELGAERIG HGVKAIEDRA LMDFLAEQQI GIESCLTSNI QTSTVAELAA HPLKTFLEHG IRASINTDDP GVQGVDIIHE YTVAAPAAGL SREQIRQAQI NGLEMAFLSA EEKRALREKV AAK | |
Greater than 90% as determined by SDS-PAGE. |
945851 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |