missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 8-oxo-dGTP diphosphatase Protein

A DNA sequence encoding the Escherichia coli (strain K12) 8-oxo-dGTP diphosphatase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7301-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16058080

  • 3391.00€ / 1 miligramo


Fecha estimada de envío
13-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_414641.1
8-oxo-dGTP diphosphatase Protein
1 mg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
944824
Greater than 90% as determined by SDS-PAGE.
Research Use Only
mutT
Recombinant Protein
Yeast
MKKLQIAVGI IRNENNEIFI TRRAADAHMA NKLEFPGGKI EMGETPEQAV VRELQEEVGI TPQHFSLFEK LEYEFPDRHI TLWFWLVERW EGEPWGKEGQ PGEWMSLVGL NADDFPPANE PVIAKLKRL
Videos
Documentos