missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S4 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8613-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_417755.1 | |
30S ribosomal protein S4 Protein | |
Research Use Only | |
rpsD | |
Recombinant Protein | |
E. coli | |
ARYLGPKLKL SRREGTDLFL KSGVRAIDTK CKIEQAPGQH GARKPRLSDY GVQLREKQKV RRIYGVLERQ FRNYYKEAAR LKGNTGENLL ALLEGRLDNV VYRMGFGATR AEARQLVSHK AIMVNGRVVN IASYQVSPND VVSIREKAKK QSRVKAALEL AEQREKPTWL EVDAGKMEGT FKRKPERSDL SADINEHLIV ELYSK | |
Greater than 90% as determined by SDS-PAGE. |
32366218 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |