missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Tight junction protein ZO-1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Tight junction protein ZO-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6798-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001003140.1 | |
Tight junction protein ZO-1 Protein | |
Research Use Only | |
TJP1 | |
Recombinant Protein | |
E. coli | |
VVATARGVFN NNGGVLSSIE TGVSIIIPQG AIPEGVEQEI YFKVCRDNSI LPPLDKEKGE TLLSPLVMCG PHGLKFLKPV ELRLPHCASM TPDGWSFALK SSDSSSGDPK TWQNKCLPGD PNYLVGANCV SVLIDHF | |
Greater than 90% as determined by SDS-PAGE. |
403752 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |