missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Serum Albumin / HSA / ALB Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5642-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916649

  • 1776.00€ / 500 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001003026.1
Serum Albumin / HSA / ALB Protein
500 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
403550
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ALB
Recombinant Protein
E. coli
EAYKSEIAHR YNDLGEEHFR GLVLVAFSQY LQQCPFEDHV KLAKEVTEFA KACAAEESGA NCDKSLHTLF GDKLCTVASL RDKYGDMADC CEKQEPDRNE CFLAHKDDNP GFPPLVAPEP DALCAAFQDN EQLFLGKYLY EIARRHPYFY APELLYYAQQ YKGVFAECCQ AADKAACLGP KIEALREKVL LSSAKERFKC ASLQKFGDRA FKAWSVARLS QRFPKADFAE ISKVVTDLTK VHKECCHGDL LECADDRADL AKYMCENQDS ISTKLKECCD KPVLEKSQCL AEVERDELPG DLPSLAADFV EDKEVCKNYQ EAKDVFLGTF LYEYARRHPE YSVSLLLRLA KEYEATLEKC CATDDPPTCY AKVLDEFKPL VDEPQNLVKT NCELFEKLGE YGFQNALLVR YTKKAPQVST PTLVEVSRKL GKVGTKCCKK PESERMSCAE DFLSVVLNRL CVLHEKTPVS ERVTKCCSES LVNRRPCFSG LEVDETYVPK EFNAETFTFH ADLCTLPEAE KQVKKQTALV ELLKHKPKAT DEQLKTVMGD FGAFVEKCCA AENKEGCFSE EGPKLVAAAQ AALV
Videos