missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Minor allergen Can f 2 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Minor allergen Can f 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7470-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16048930

  • 1857.00€ / 1 miligramo


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001003189.1
Minor allergen Can f 2 Protein
1 mg
Not determined.
Full Length
Dog
His-SUMO
Tris-based buffer, 50% glycerol
403829
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CANF2
Recombinant Protein
E. coli
QEGNHEEPQG GLEELSGRWH SVALASNKSD LIKPWGHFRV FIHSMSAKDG NLHGDILIPQ DGQCEKVSLT AFKTATSNKF DLEYWGHNDL YLAEVDPKSY LILYMINQYN DDTSLVAHLM VRDLSRQQDF LPAFESVCED IGLHKDQIVV LSDDDRCQGS RD
Videos
Documentos