missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog CMA1 / Chymase 1 Protein

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5843-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918189

  • 530.00€ / 50 microgramos


Fecha estimada de envío
23-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001013442.1
CMA1 / Chymase 1 Protein
50 μg
Not determined.
Full Length
Dog
His
Tris-based buffer, 50% glycerol
490628
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CMA1
Recombinant Protein
Yeast
IIGGTESKPH SRPYMAHLEI LTLRNHLASC GGFLIRRNFV LTAAHCAGRF IMVTLGAHNI QKKEDTWQKL EVIKQFPHPK YDDLTLRHDI MLLKLKEKAN LTLAVGTLPL SPQFNFVPPG RMCRVAGWGK RQVNGSGSDT LQEVKLRLMD PQACRHYMAF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGQ NDAKPPAVFT RISHYRPWIN KVLKQNKA
Videos