missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus, ST4 Dengue NS1 Protein

A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10497-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914058

  • 1372.00€ / 1 miligramo


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Dengue Virus, ST4 Dengue NS1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus, ST4
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline and 50mM arginine.
Videos