missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Premembrane Protein
A cDNA sequence encoding the Dengue Premembrane was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11652-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Dengue Premembrane Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM phosphate buffer saline, pH 7.4. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |