missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Premembrane Protein

A cDNA sequence encoding the Dengue Premembrane was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP11652-1mg

Weitere Details : Peso : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15907928

  • 1815.00€ / 1 miligramo
Fecha estimada de envío: 12-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Beschreibung

Beschreibung

Spezifikation

Spezifikation

Dengue Premembrane Protein
Research Use Only
Recombinant Protein
E. coli
50mM phosphate buffer saline, pH 7.4.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote