missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue NS1 ST4 Protein

A cDNA sequence encoding the Dengue NS1 ST4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11647-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937908

  • 5660.00€ / 1 miligramo


Fecha estimada de envío
17-08-2020
Añadir a la cesta

Especificaciones

Especificaciones

Dengue NS1 ST4 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
Untagged
Protein is >95% pure as determined by 12.5% SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
Dengue NS1 ST4 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1?g/ml of Leupeptin, Aprotinin and Pepstatin A.
Videos
Documentos