missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue NS1 n Protein
A cDNA sequence encoding the Dengue NS1 n was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11643-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Dengue NS1 n Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
100mM Tris-Hcl pH 8.0, 60mM NaCl, and 5% Glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
GST | |
Protein is >90% pure as determined by SDS-PAGE. |