missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Cytomegalovirus Viral interleukin-10 homolog Protein

A DNA sequence encoding the Human cytomegalovirus (strain Merlin) (HHV-5) (Human herpesvirus 5) Viral interleukin-10 homolog, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7461-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16048920

  • 1487.00€ / 500 microgramos


Fecha estimada de envío
23-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

YP_081552.1
Viral interleukin-10 homolog Protein
500 μg
Not determined.
Full Length of Mature. Protein
Cytomegalovirus
Untagged
Tris-based buffer, 50% glycerol
3077506
Greater than 95.8% as determined by SDS-PAGE.
Research Use Only
UL111A
Recombinant Protein
E. coli
ATTTTIKNTK PQCRPEDYAT RLQDLRVTFH RVKPTLQRED DYSVWLDGTV VKGCWGCSVM DWLLRRYLEI VFPAGDHVYP GLKTELHSMR STLESIYkDa MRQCPLLGCG DKSVISRLSQ EAERKSDNGT RKGLSELDTL FSRLEEYLHS RK
Videos
Documentos