Learn More
enQuireBio™ Recombinant Cytomegalovirus Viral interleukin-10 homolog Protein
A DNA sequence encoding the Human cytomegalovirus (strain AD169) (HHV-5) (Human herpesvirus 5) Viral interleukin-10 homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7080-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Viral interleukin-10 homolog Protein | |
Research Use Only | |
UL111A | |
Recombinant Protein | |
E. coli | |
SEEAKPATTT IKNTKPQCRP EDYATRLQDL RVTFHRVKPT LQREDDYSVW LDGTVVKGCW GCSVMDWLLR RYLEIVFPAG DHVYPGLKTE LHSMRSTLES IYKDMRQCPL LGCGDKSVIS RLSQEAERKS DNGTRKGLSE LDTLFSRLEE YLHSRK | |
Greater than 90% as determined by SDS-PAGE. |
500 μg | |
Not determined. | |
Full Length | |
Cytomegalovirus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |