missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chinese Hamster P4HB / ERBA2L Protein

A DNA sequence encoding the Chinese hamster P4HB / ERBA2L, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7857-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051221

  • 530.00€ / 50 microgramos


Fecha estimada de envío
22-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001233622.1
P4HB / ERBA2L Protein
50 μg
Not determined.
Full Length
Chinese Hamster
His
Tris-based buffer, 50% glycerol
100689433
Greater than 90% as determined by SDS-PAGE.
Research Use Only
P4hb
Recombinant Protein
Yeast
DAPEEEDNVL VLKKSNFAEA LAAHNYLLVE FYAPWCGHCK ALAPEYAKAA AKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFKN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLSDTAAAE TLIDSSEVAV IGFFKDVESD SAKQFLLAAE AVDDIPFGIT SNSGVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKEKLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLGNFKK AAEGFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESDELTAE KITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EEVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIIIAKMDS TANEVEAVKV HSFPTLKFFP ATADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDVDLE EALEPDMEED DDQKAVKDEL
Videos
Documentos