Learn More
enQuireBio™ Recombinant Chikungunya Virus VLP Protein
Recombinant Protein
Marca: enQuireBio™ QP10473-500ug
Detalles adicionales : Peso : 0.01000kg
Descripción
Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.Especificaciones
VLP Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
0.02M tris-HCl pH7.5, 0.125M sodium chloride. |
500 μg | |
Untested | |
Chikungunya virus (Strain: Senegal/37997) | |
Native | |
>90% as determined by SDS-PAGE. |