Learn More
enQuireBio™ Recombinant Chikungunya Virus VLP Protein
Recombinant Protein
Marca:  enQuireBio™ QP10473-500ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15913988
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Descripción
Descripción
Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Especificaciones
Especificaciones
VLP Protein
Research Use Only
Recombinant Protein
Human
0.02M tris-HCl pH7.5, 0.125M sodium chloride.
500 μg
Untested
Chikungunya virus (Strain: Senegal/37997)
Native
>90% as determined by SDS-PAGE.