missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Mitochondrial Rho GTPase 1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Mitochondrial Rho GTPase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7700-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001006208.1 | |
Mitochondrial Rho GTPase 1 Protein | |
Research Use Only | |
RHOT1 | |
Recombinant Protein | |
E. coli | |
MKKDVRILLV GEPRVGKTSL IMSLVSEEFP EEVPPRAEEI TIPADVTPER VPTHIVDYSE AEQNDEQLYH EISQANVICI VYAVNNKNSI DKVTSRWIPL INERTDKDSR LPLILVGNKS DLVEYSSMET ILPIMNQYTE IETCVECSAK NLKNRSELFY YAQKAVLHPT GPLYCPEEKE MKPACIKALT RIFRISDQDN DGTLNDAELN FFQRICFNT | |
Greater than 90% as determined by SDS-PAGE. |
417410 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His-SUMO | |
Tris-based buffer, 50% glycerol |