missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken GH Antagonist Protein

A cDNA sequence encoding the GH Antagonist was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10641-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15924498

  • 1285.00€ / 1 miligramo
Fecha estimada de envío: 29-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

GH Antagonist Protein
Research Use Only
Recombinant Chicken Growth Hormone G119R mutant did not bind to ovine GHR-ECD and was devoid of any biological activity in FDC-P1 3B9 cells. However, in binding experiments that were carried out using chicken liver membranes, both ovine GH and chicken GH showed similar IC50 values in competition with 125I-ovine GH, while the IC50 of G119R mutein was 10-fold higher. These results emphasize the importance of species specificity and indicate the possibility of antagonistic activity of chGH G119R in homologous system.
Chicken
Untagged
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 pH 8.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales