missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken GAPDH Protein

A DNA sequence encoding the Gallus gallus (Chicken) GAPDH, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6069-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919759

  • 1337.00€ / 200 microgramos


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_989636.1
GAPDH Protein
200 μg
Not determined.
Full Length
Chicken
His
Tris-based buffer, 50% glycerol
374193
Greater than 90% as determined by SDS-PAGE.
Research Use Only
GAPDH
Recombinant Protein
E. coli
VKVGVNGFGR IGRLVTRAAV LSGKVQVVAI NDPFIDLNYM VYMFKYDSTH GHFKGTVKAE NGKLVINGHA ITIFQERDPS NIKWADAGAE YVVESTGVFT TMEKAGAHLK GGAKRVIISA PSADAPMFVM GVNHEKYDKS LKIVSNASCT TNCLAPLAKV IHDNFGIVEG LMTTVHAITA TQKTVDGPSG KLWRDGRGAA QNIIPASTGA AKAVGKVIPE LNGKLTGMAF RVPTPNVSVV DLTCRLEKPA KYDDIKRVVK AAADGPLKGI LGYTEDQVVS CDFNGDSHSS TFDAGAGIAL NDHFVKLVSW YDNEFGYSNR VVDLMVHMAS KE
Videos