missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chicken Anosmin-1 Protein
A DNA sequence encoding the Gallus gallus (Chicken) Anosmin-1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6250-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_990755.1 | |
Anosmin-1 Protein | |
Research Use Only | |
ANOS1 | |
Recombinant Protein | |
E. coli | |
SPAGPGAATA RRQDEAFSTA RCTSRCLSLQ ITRISAFFKH FQNNGSLAWC QNHKQCSKCL EPCKESWDLK KNHCQSFCEP LFPKKNYECL TSCEFLKYIL SVKQGDCPAP EKASGFAAAC VESCEADSEC SGVKKCCSNG CGHTCQVPKN LYKGVPLKPR KELKFIELQS GDLEVKWSSK FNISIEPVIY VVQRRWNQGI HPSEDDATNW QTVAQTTDER VQLSDIRASR WYQFRVAAVN VHGTRGFTAP SKHFRSSKDP | |
Greater than 90% as determined by SDS-PAGE. |
396395 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chicken | |
His | |
Tris-based buffer, 50% glycerol |