missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine MCP 2 Protein
A cDNA sequence encoding the MCP 2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10782-1mg
Additional Details : Peso : 0.01000kg
Specifications
448792 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-100 ng/ml. | |
Canine | |
Untagged | |
MCP-2 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
MCP 2 Protein | |
Research Use Only | |
CCL8 | |
Recombinant Protein | |
E. coli | |
QPDSVSIPIT CCFSMVKRKI PMQKLESYMR ITNSQCPQEA VIFKTKASRE ICADPKQKWV QDYMNHLDQK SQAQKP | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |