missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine CASQ2 Protein
A cDNA sequence encoding the CASQ2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11280-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
483134 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Canine | |
MKRTHLFIAG LYLLASCRAE EGLNFPTYDG KDRVVSLTEK NFKQVLKKYD VLCLYYHESV SSDKVAQKQF QLKEIVLELV AQVLEHKDIG FVMVDAKKEA KLAKKLGFDE EGSLYVLKGD RTIEFDGEFA ADVLVEFLLD LIEDPVEIIN SKLEVQAFER IEDQIKLIGF FKSEESEYYK AFEEAAEHFQ PYIKFFATFD KGVAKKLSLK MNEVDFYEPF MDEPIAIPDK PYTEEELVEF VKEHQRPTLR RLRPEDMFET WEDDLNGIHI VAFAERSDPD GYEFLEILKQ VARDNTDNPD LSIVWIDPDD FPLLVAYWEK TFKIDLFKPQ IGVVNVTDAD SVWMEIPDDD DLPTAEELED WIEDVLSGKI NTEDDDNEEG DDGDDDEDDD DDDGNNSDEE SNDDSDDDDE | |
Greater than 90% as determined by SDS-PAGE. |
CASQ2 Protein | |
Research Use Only | |
CASQ2 | |
Canine | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 10mM Tris-HCl and 1mM EGTA. |