Learn More
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin, serovar A Protein
A DNA sequence encoding the Chlamydia trachomatis Major outer membrane porin, serovar A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7129-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Major outer membrane porin, serovar A Protein | |
1 mg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLKTDVNKEF QMGAAPTTRD VAGLEKDPVV NVARPNPAYG KHMQDAEMFT NAAYMALNIW DRFDVFCTLG ATTGYLKGNS ASFNLVGLFG TKTQSSGFDT ANIVPNTALN QAVVELYTDT TFAWSVGARA ALWECGCATL GASFQYAQSK PKVEELNVLC NASEFTINKP KGYVGAEFPL DITAGTEAAT GTKDASIDYH EWQASLALSY RLNMFTPYIG VKWSRVSFDA DTIRIAQPKL AKPVLDTTTL NPTIAGKGTV VSSAENELAD TMQIVSLQLN KMKSRKSCGI AVGTTIVDAD KYAVTVETRL IDERAAHVNA QFRF |