missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. elegans SNAP25 Protein

A cDNA sequence encoding the SNAP25 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13532-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913879

  • 103.00€ / 10 microgramos


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

SNAP25 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
C. elegans
Untagged
The protein contains 20mM Tris-HCl pH 7.5, 50mM NaCl, 5mM DTT, 1mM EDTA and 10% Glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MSGDDDIPEG LEAINLKMNA TTDDSLESTR RMLALCEESK EAGIKTLVML DDQGEQLERCEGALDTINQD MKEAEDHLKG MEKCCGLCVL PWNKTDDFEK TEFAKAWKKD DDGGVISDQPRITVGDSSMG PQGGYITKIT NDAREDEMDE NVQQVSTMVG NLRNMAIDMS TEVSNQNRQL DRIHDKAQSN EVRVESANKR AKNLITK
Videos