missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type C1 Protein
A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type C1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7104-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Botulinum neurotoxin type C1 Protein | |
500 μg | |
Not determined. | |
Recombinant Protein | |
E. coli | |
PITINNFNYS DPVDNKNILY LDTHLNTLAN EPEKAFRITG NIWVIPDRFS RNSNPNLNKP PRVTSPKSGY YDPNYLSTDS DKDPFLKEII KLFKRINSRE IGEELIYRLS TDIPFPGNNN TPINTFDFDV DFNSVDVKTR QGNNWVKTGS INPSVIITGP RENIIDPETS TFKLTNNTFA AQEGFGALSI ISISPRFMLT YSNATNDVGE GRFSKSEFCM DPILILMHEL NHAMHNLYGI AIPNDQTISS VTSNIFYSQY NVKLEYAEIY AFGGPTIDLI PKSARKYFEE KALDYYRSIA KRLNSITTAN PSSFNKYIGE YKQKLIRKYR FVVESSGEVT VNRNKFVELY NELTQIFTEF NYAKIYNVQN RKIYLSNVYT PVTANILDDN VYDIQNGFNI PKSNLNVLFM GQNLSRNPAL RKVNPEN |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Partial (see sequence information for more details). | |
C. botulinum | |
His-SUMO | |
Tris-based buffer, 50% glycerol |