missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Fibronectin Protein

A cDNA sequence encoding the Fibronectin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11880-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918678

  • 160.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 04-04-2019
Añadir a la cesta

Especificación

Especificación

Fibronectin Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Native Protein
The Bovine Fibronectin was lyophilized from a concentrated 1 mg/ml solution containing 0.05M Tris Cl pH 7.5 and 0.1M NaCl.
Videos