missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Erythropoietin / EPO Protein
A DNA sequence encoding the Bos taurus (Bovine) Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5987-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_776334.1 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
Recombinant Protein | |
Yeast | |
APARLICDSR VLERYILEAR EAENATMGCA EGCSFNENIT VPDTKVNFYA WKRMEVQQQA LEVWQGLALL SEAILRGQAL LANASQPCEA LRLHVDKAVS GLRSLTSLLR ALGAQKEAIS LPDATPSAAP LRAFTVDALS KLFRIYSNFL RGKLTLYTGE ACRRGDR | |
Greater than 90% as determined by SDS-PAGE. |
280784 | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |