missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine CD8A / MAL Protein

A DNA sequence encoding the Bos taurus (Bovine) CD8A / MAL, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5800-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15907869

  • 3391.00€ / 1 miligramo


Fecha estimada de envío
26-06-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_776440.1
CD8A / MAL Protein
1 mg
Not determined.
Extracellular Domain
Bovine
His
Tris-based buffer, 50% glycerol
281060
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD8A
Recombinant Protein
Yeast
LSFRMSPTQK ETRLGEKVEL QCELLQSGMA TGCSWLRHIP GDDPRPTFLM YLSAQRVKLA EGLDPRHISG AKVSGTKFQL TLSSFLQEDQ GYYFCSVVSN SILYFSNFVP VFLPAKPATT PAMRPSSAAP TSAPQTRSVS PRSEVCRTSA GSAVDTSRLD FACN
Videos