missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine CCL20 / MIP-3 alpha Protein
A DNA sequence encoding the Bos taurus (Bovine) CCL20 / MIP-3 alpha, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7906-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_776688.1 | |
CCL20 / MIP-3 alpha Protein | |
Research Use Only | |
CCL20 | |
Recombinant Protein | |
Yeast | |
ASNFDCCLRY TERILHPSIL VGFTQQLANE ACDINAVVFY TRKKLAVCAD PKKKWVKQVV HMLSQRVKRM | |
Greater than 90% as determined by SDS-PAGE. |
281666 | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |