missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine APOH Protein
A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11053-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
281006 | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Bovine | |
MPPPALVLLL GFLCHVAIAG RTCPKPDELP FSTVVPLKRT YEPGEQIVFS CQPGYVSRGG IRRFTCPLTG LWPINTLKCM PRVCPFAGIL ENGTVRYTTF EYPNTISFSC HTGFYLKGAS SAKCTEEGKW SPDLPVCAPI TCPPPPIPKF ASLSVYKPLA GNNSFYGSKA VFKCLPHHAM FGNDTVTCTE HGNWTQLPEC REVRCPFPSR PDNGFVNHPA NPVLYYKDTA TFGCHETYSL DGPEEVECSK FGNWSAQPSC KASCKLSIKR ATVIYEGERV AIQNKFKNGM LHGQKVSFFC KHKEKKCSYT EDAQCIDGTI EIPKCFKEHS SLAFWKTDAS DVKPC | |
Greater than 70% as determined by SDS-PAGE. |
APOH Protein | |
Research Use Only | |
APOH | |
Bovine | |
Untagged | |
APOH protein solution is supplied in 16mM Tris-HCl pH 8.0, 280mM NaCl, 0.16mM AEBSF and 20% glycerol. |