missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine APOH Protein

A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11053-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915998

  • 103.00€ / 10 microgramos


Fecha estimada de envío
17-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

281006
Greater than 70% as determined by SDS-PAGE.
Research Use Only
APOH
Bovine
Untagged
APOH protein solution is supplied in 16mM Tris-HCl pH 8.0, 280mM NaCl, 0.16mM AEBSF and 20% glycerol.
APOH Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Native Protein
MPPPALVLLL GFLCHVAIAG RTCPKPDELP FSTVVPLKRT YEPGEQIVFS CQPGYVSRGG IRRFTCPLTG LWPINTLKCM PRVCPFAGIL ENGTVRYTTF EYPNTISFSC HTGFYLKGAS SAKCTEEGKW SPDLPVCAPI TCPPPPIPKF ASLSVYKPLA GNNSFYGSKA VFKCLPHHAM FGNDTVTCTE HGNWTQLPEC REVRCPFPSR PDNGFVNHPA NPVLYYKDTA TFGCHETYSL DGPEEVECSK FGNWSAQPSC KASCKLSIKR ATVIYEGERV AIQNKFKNGM LHGQKVSFFC KHKEKKCSYT EDAQCIDGTI EIPKCFKEHS SLAFWKTDAS DVKPC
Videos