missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Adiponectin / Acrp30 / ADIPOQ Protein

A DNA sequence encoding the Bos taurus (Bovine) Adiponectin / Acrp30 / ADIPOQ, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7647-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050001

  • 491.00€ / 50 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_777167.1
Adiponectin / Acrp30 / ADIPOQ Protein
50 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
282865
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ADIPOQ
Recombinant Protein
Yeast
EDNMEDPPLP KGACAGWMAG IPGHPGHNGT PGRDGRDGTP GEKGEKGDPG LVGPKGDTGE TGITGIEGPR GFPGTPGRKG EPGESAYVYR SAFSVGLERQ VTVPNVPIRF TKIFYNQQNH YDGTTGKFLC NIPGLYYFSY HITVYLKDVK VSLYKNDKAL LFTHDQFQDK NVDQASGSVL LYLEKGDQVW LQVYEGENHN GVYADNVNDS TFTGFLLYHN IVE
Videos
Documentos