missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. baumannii Elongation factor Tu Protein

A DNA sequence encoding the Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377) Elongation factor Tu, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6854-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056310

  • 352.00€ / 10 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

Elongation factor Tu Protein
10 μg
Not determined.
Full Length
A. baumannii
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
tuf1
Recombinant Protein
Yeast
MAKAKFERNK PHVNVGTIGH VDHGKTTLTA AIATICAKTY GGEAKDYSQI DSAPEEKARG ITINTSHVEY DSPTRHYAHV DCPGHADYVK NMITGAAQMD GAILVCAATD GPMPQTREHI LLSRQVGVPY IIVFLNKCDL VDDEELLELV EMEVRELLST YDFPGDDTPV IRGSALAALN GEAGPYGEES VLALVAALDS YIPEPERAID KAFLMPIEDV FSISGRGTVV TGRVEAGIIK VGEEVEIVGI KDTVKTTVTG VEMFRKLLDE GRAGENCGIL LRGTKREEVQ RGQVLAKPGT IKPHTKFDAE VYVLSKEEGG RHTPFLNGYR PQFYFRTTDV TGAIQLKEGV EMVMPGDNVE MSVELIHPIA MDPGLRFAIR EGGRTVGAGV VAKVTA
Videos
Documentos