missing translation for 'onlineSavingsMsg'
Learn More

RCC2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 16755-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816494

  • 109.00€ / 20 microlitros

Envío estimado: 21-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations.
Spécification

Spécification

RCC2
0.18 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9P258
KIAA1470, Protein RCC2, RCC1 like protein TD 60, RCC2, TD 60, TD60
Rabbit
IgG
20 μL
-20°C
Primary
55920
Western Blot
Unconjugated
RCC2
Liquid
RCC2
RCC2 Fusion Protein Ag10271
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents
Certificats