missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAS1 Mouse anti-Human, Clone: 4H8A12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66170-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5′-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.Especificaciones
RCAS1 | |
Monoclonal | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00559 | |
EBAG9 | |
RCAS1 Fusion Protein Ag2905 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
4H8A12 | |
Unconjugated | |
EBAG9 | |
EB9, EBAG9, PDAF, RCAS1 | |
Mouse | |
Protein G | |
RUO | |
9166 | |
-20°C | |
Liquid |