missing translation for 'onlineSavingsMsg'
Learn More

RCAN1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-46852AF350

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214344

  • 561.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

RCAN1 Polyclonal specifically detects RCAN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

RCAN1
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1
Rabbit
Affinity Purified
Cancer, Neuroscience, Signal Transduction
1827.0
Human, Mouse
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
RCAN1
Synthetic peptide made to an internal portion of human RCAN1 (within residues 40-90). [Swiss-ProtNo. P53805]
0.1 mL
Primary
This antibody is specific for isoform 1-1L.
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales