missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPJ Mouse anti-Human, Clone: 2F12G12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66132-1-IG-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Transcriptional regulator that plays a central role in Notch signaling, a signaling pathway involved in cell-cell communication that regulates a broad spectrum of cell-fate. determinations. Acts as a transcriptional repressor when it is not associated with Notch proteins. When associated with some Notch protein, it acts as a transcriptional activator that activates transcription of Notch target genes. Probably represses or activates transcription via the recruitment of chromatin remodeling complexes containing histone deacetylase or histone acetylase proteins, respectively. Specifically binds to the immunoglobulin kappa-type J segment recombination signal sequence.Especificaciones
RBPJ | |
Monoclonal | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q06330 | |
RBPJ | |
RBPJ Fusion Protein Ag6302 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
2F12G12 | |
Unconjugated | |
RBPJ | |
CBF 1, CBF1, csl, IGKJRB, IGKJRB1, KBF2, NY-REN-30, RBP J, RBP J kappa, RBP JK, RBPJ, RBPJK, RBPSUH, SUH | |
Mouse | |
Protein G | |
RUO | |
3516 | |
-20°C | |
Liquid |