Learn More
Novus Biologicals™ Rat Factor XIII ELISA Kit (Colorimetric)
Rat Factor XIII ELISA Kit (Colorimetric) measures Rat Factor XIII in Serum, plasma, cell supernatant, and other biological fluids.
Marca:  Novus Biologicals™ NBP2-82475
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213449
  • 769.96€ / 1 set
Fecha estimada de envío: 14-08-2024
para ver el stock
Especificaciones
0.63 to 40 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat Factor XIII in samples. No Significant cross-reactivity or interference between Rat Factor XIII and analogues was observed.
ELISA
F13B
Storage of components varies. See protocol for specific instructions.
Factor XIII
0.38 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Rat
2165
1 Kit
bA525O21.1 (coagulation factor XIII, A1 polypeptide), coagulation factor XIII A chain, coagulation factor XIII, A1 polypeptide, Coagulation factor XIIIa, EC 2.3.2.13, F13Acoagulation factor XIII, A polypeptide, factor XIIIa, fibrin stabilizing factor, A subunit, fibrinoligase, FSF, A subunit, Protein-glutamine gamma-glutamyltransferase A chain, TGase, Transglutaminase A chain, transglutaminase. plasma