Learn More
Novus Biologicals™ Rat Calnexin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Marca:  Novus Biologicals™ NBP2-75818
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213889
  • 799.12€ / 1 set
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
The Rat Calnexin ELISA Kit (Colorimetric) measures rat Calnexin in serum, plasma and other biological fluids.
Especificaciones
Especificaciones
0.16 - 10 ng/mL
This kit recognizes Rat CNX in samples. No significant cross-reactivity or interference between Rat CNX and analogues was observed.
Biological Fluids, Plasma, Serum
821
Rat
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
CANX
1 Kit
Calnexin