Learn More
Invitrogen™ RARG Recombinant Human Protein, Ligand Binding Domain, GST-Tagged
Retinoic acid receptor gamma, ligand binding domain (RARG-LBD) is a 58.85 kDa recombinant human protein (amino acids 178-454) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells.
Marca: Invitrogen™ PV4413
Detalles adicionales : Peso : 0.50000kg
Descripción
RARG-LBD is expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator recruitment studies.
Especificaciones
Nuclear Receptor Assay | |
50 μg | |
RUO - research use only | |
Retinoic Acid Receptor gamma | |
Recombinant | |
Hormone Receptors |
5916 | |
Store in ultra-cold freezer (-68 to -85°C). | |
Recombinant, Ligand Binding Domain | |
Human | |
GST Tag |